Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-Piperidine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific
Synonyms: D-Pipecolinic AcidD-Homoproline(R)-(+)-2-Piperidinecarboxylic AcidH-D-HoPro-OHH-D-Homopro-OHD(+)-Pipecolinic acid

075001-5G 075001-1G 075001-10G
102539-262EA 61.46 USD InStock
102539-262 102539-264 102539-266
(R)-Piperidine-2-carboxylic acid ≥95%
(R)-Piperidine-2-carboxylic acid
Hazard: Irritant


Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00064346
CAS Number: 1723-00-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty