Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocyclopentane ≥95%

Supplier: Matrix Scientific
Synonyms: Cyclopentyl bromide

075015-10G 075015-100G
102539-330EA 301.94 USD InStock
102539-330 102539-332
Bromocyclopentane ≥95%
Bromocyclopentane
Hazard: Flammable


Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 137…139 °C
Density: 1.39
Flash Pt: 42 °C
MDL Number: MFCD00001359
CAS Number: 137-43-9
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty