Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzothiazole ≥95%

Supplier: Matrix Scientific

075058-100G 075058-5G 075058-10G
102539-542EA 126.15 USD InStock
102539-542 102539-544 102539-546
2-Chlorobenzothiazole ≥95%
2-Chlorobenzothiazole
Hazard: Irritant


Formula: C₇H₄ClNS
MW: 169.63 g/mol
MDL Number: MFCD00005776
CAS Number: 615-20-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty