Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Bromophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

075124-100G
102539-884EA 169.23 USD InStock
102539-884
(2-Bromophenyl)acetonitrile ≥95%
(2-Bromophenyl)acetonitrile
Hazard: Toxic/Lachrymator


Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 252 °C
Melting Pt: 19 °C
Density: 1.51
Flash Pt: 113 °C
MDL Number: MFCD00001896
CAS Number: 19472-74-3
UN: 1694
ADR: 6.1,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty