Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Fluoro-4-nitrobenzene ≥95%

Supplier: Matrix Scientific
Synonyms: 4-Fluoronitrobenzene

075181-100G
102540-180EA 53.85 USD InStock
102540-180
1-Fluoro-4-nitrobenzene ≥95%
1-Fluoro-4-nitrobenzene
Hazard: Toxic


Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 205 °C
Melting Pt: 22…24 °C
MDL Number: MFCD00007282
CAS Number: 350-46-9
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty