Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-5-bromovalerate ≥95%

Supplier: Matrix Scientific
Synonyms: Ethyl 5-bromovalerate5-Bromovaleric acid ethyl ester

075188-100G 075188-10G
102540-216EA 463.08 USD InStock
102540-216 102540-214
Ethyl-5-bromovalerate ≥95%
Ethyl 5-bromovalerate
Hazard: Irritant


Formula: BrH₂C(CH₂)₃COOCH₂CH₃
MW: 209.08 g/mol
MDL Number: MFCD00000266
CAS Number: 14660-52-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty