Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-cysteine ≥95%

Supplier: Matrix Scientific
Synonyms: Acetylcysteine(R)-2-Acetamido-3-mercaptopropanoic acid(R)-(+)-N-AcetylcysteineAc-Cys-OH

075227-100G
102540-400EA 76.92 USD InStock
102540-400
N-Acetyl-L-cysteine ≥95%
N-Acetyl-L-cysteine
Hazard: Irritant


Formula: C₅H₉NO₃S
MW: 163.2 g/mol
Melting Pt: 106…111 °C
MDL Number: MFCD00004880
CAS Number: 616-91-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty