Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-2-cyclohexylacetic acid ≥95%

Supplier: Matrix Scientific
Synonyms: H-Chg-OH(S)-alpha-Aminocyclohexylacetic AcidH-Cyclohexyl-Gly-OH

075299-100G 075299-10G 075299-5G
102540-736EA 340 USD InStock
102540-736 102540-734 102540-732
(S)-2-Amino-2-cyclohexylacetic acid ≥95%
(S)-2-Amino-2-cyclohexylacetic acid
Hazard: Irritant


Formula: C₈H₁₅NO₂
MW: 157.2 g/mol
MDL Number: MFCD01311679
CAS Number: 14328-51-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty