Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Formylbenzoic acid ≥95%

Supplier: Matrix Scientific
Synonyms: 2-CarboxybenzaldehydePhthalaldehydic acid

075300-100G 075300-10G
102540-738EA 240.79 USD InStock
102540-738 102540-740
2-Formylbenzoic acid ≥95%
2-Formylbenzoic acid
Hazard: Irritant


Formula: C₈H₆O₃
MW: 150.13 g/mol
MDL Number: MFCD00003336
CAS Number: 119-67-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty