Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzonitrile ≥95%

Supplier: Matrix Scientific
Synonyms: m-Bromobenzonitrile

075372-100G 075372-25G
102541-058EA 176.92 USD InStock
102541-058 102541-056
3-Bromobenzonitrile ≥95%
3-Bromobenzonitrile
Hazard: Irritant-Harmful


Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 224…226 °C
Melting Pt: 37…41 °C
MDL Number: MFCD00001796
CAS Number: 6952-59-6
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty