Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Tryptophan ≥95%

Supplier: Matrix Scientific
Synonyms: H-D-Trp-OH

075483-100G
102541-564EA 135.38 USD InStock
102541-564
D-(+)-Tryptophan ≥95%
D(+)-Tryptophan
Hazard: Irritant


Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
MDL Number: MFCD00005647
CAS Number: 153-94-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty