Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Aspartic acid ≥95%

Supplier: Matrix Scientific
Synonyms: (R)-(-)-2-Amino-1,4-butanedioic acid(R)-(-)-Aspartic acidD(-)-2-Amino-1,4-butanedioic acidH-D-Asp-OH(R)-2-Aminosuccinic acid

075484-100G
102541-566EA 138.46 USD InStock
102541-566
D-(-)-Aspartic acid ≥95%
D(-)-Aspartic acid
Hazard: Irritant


Formula: C₄H₇NO₄
MW: 133.1 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00063081
CAS Number: 1783-96-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty