Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitrophthalonitrile ≥95%

Supplier: Matrix Scientific
Synonyms: 3-Nitrobenzene-1,2-dicarbonitrile3-Nitro-1,2-dicyanobenzene3-Nitro-1,2-benzenedicarbonitrile

075532-100G
102541-784EA 113.85 USD InStock
102541-784
3-Nitrophthalonitrile ≥95%
3-Nitrophthalonitrile
Hazard: Irritant


Formula: C₈H₃N₃O₂
Melting Pt: 162…165 °C
MDL Number: MFCD00191558
CAS Number: 51762-67-5
UN: 3276
ADR: 6.1,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty