Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-Methyl-2-amino-2-phenylacetate hydrochloride ≥95%

Supplier: Matrix Scientific
Synonyms: H-D-Phg-OMe.HClD-(-)-2-Phenylglycine methyll ester hydrochloride(R)-2-Phenylglycine methyl ester hydrochlorideMethyl (R)-(-)-2-Amino-2-phenylacetate hydrochloride(R)-(-)-2-Amino-2-phenylacetic acid methyll ester hydrochlorideH-D-Phg-OMe·HCl

075544-100G
102541-838EA 249.23 USD InStock
102541-838
(R)-Methyl-2-amino-2-phenylacetate hydrochloride ≥95%
(R)-Methyl-2-amino-2-phenylacetate hydrochloride
Hazard: Irritant


Formula: C₉H₁₁NO₂·HCl
MW: 201.7 g/mol
MDL Number: MFCD00137487
CAS Number: 19883-41-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty