Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride ≥95%

Supplier: Matrix Scientific
Synonyms: H-D-Phe-OtBu.HClD-Phenylalanine tert-butyl ester hydrochlorideH-D-Phe-OtBu·HCl

075724-10G 075724-5G 075724-1G
102542-722EA 440 USD InStock
102542-722 102542-720 102542-718
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride ≥95%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride
Hazard: Irritant


Formula: C₁₃H₁₉NO₂·HCl
MW: 257.8 g/mol
MDL Number: MFCD00153455
CAS Number: 3403-25-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty