Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrofluorene ≥95%

Supplier: Matrix Scientific
Synonyms: 2-Nitro-9H-fluorene

075927-100G 075927-5G 075927-10G
102543-668EA 86.15 USD InStock
102543-668 102543-670 102543-672
2-Nitrofluorene ≥95%
2-Nitrofluorene
Hazard: Irritant


Formula: C₁₃H₉NO₂
MW: 211.22 g/mol
MDL Number: MFCD00001117
CAS Number: 607-57-8
UN: 3077
ADR: 9,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty