Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,N-Trimethylanilinium chloride ≥95%

Supplier: Matrix Scientific
Synonyms: Trimethylphenylammonium chloridePhenyltrimethylammonium chloride

075946-10G 075946-100G
102543-754EA 36.92 USD InStock
102543-754 102543-756
N,N,N-Trimethylanilinium chloride ≥95%
N,N,N-Trimethylanilinium chloride
Hazard: Irritant


Formula: C₉H₁₄ClN
MW: 171.67 g/mol
MDL Number: MFCD00011790
CAS Number: 138-24-9
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty