Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mefenamic acid ≥95%

Supplier: Matrix Scientific
Synonyms: 2-((2,3-Dimethylphenyl)amino)benzoic acid2-(2,3-Dimethylphenylamino)benzoic acid2-(2,3-Xylidino)benzoic acidN-(2,3-Dimethylphenyl)anthranilic acid

076071-100G 076071-5G 076071-10G
102544-352EA 235.38 USD InStock
102544-352 102544-350 102544-348
Mefenamic acid ≥95%
Mefenamic acid
Purity: 95+%
CAS: 61-68-7
MDL Number: MFCD00051721
Molecular Formula: C15H15NO2
Molecular Weight: 241,29
Hazard: Irritant


Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
MDL Number: MFCD00051721
CAS Number: 61-68-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty