Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Lysine monohydrochloride ≥95%

Supplier: Matrix Scientific
Synonyms: H-DL-Lys-OH · HCl(±)-2,6-Diaminohexanoic acid monohydrochlorideH-DL-Lys-OH.HCl(±)-2,6-Diaminocaproic acid monohydrochloride(±)-Lysine monohydrochloride

076147-100G 076147-10G
102544-706EA 155.38 USD InStock
102544-706 102544-704
DL-Lysine monohydrochloride ≥95%
DL-Lysine monohydrochloride
Hazard: Irritant


Formula: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
MDL Number: MFCD00064563
CAS Number: 70-53-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty