Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Methionine ≥95%

Supplier: Matrix Scientific
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acidL(-)-MethionineMethionine(S)-(-)-2-Amino-4-(methylthio)butanoic acidH-Met-OH(S)-(-)-Methionine(S)-2-Amino-4-(methylthio)butanoic acid

076155-100G
102544-740EA 78.46 USD InStock
102544-740
L-Methionine ≥95%
L-Methionine
Hazard: Irritant


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD00063097
CAS Number: 63-68-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty