Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-2,4,6-triiodoisophthalic acid ≥95%

Supplier: Matrix Scientific

076231-100G
102545-110EA 83.08 USD InStock
102545-110
5-Amino-2,4,6-triiodoisophthalic acid ≥95%
5-Amino-2,4,6-triiodoisophthalic acid
Hazard: Irritant


Formula: C₈H₄I₃NO₄
MW: 558.84 g/mol
MDL Number: MFCD00190167
CAS Number: 35453-19-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty