Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2'-Bipyridyl ≥95%

Supplier: Matrix Scientific
Synonyms: 2,2'-Bipyridine2,2'-Dipyridylα,α′-Dipyridyl

076381-100G
102545-880EA 75.38 USD InStock
102545-880
2,2'-Bipyridyl ≥95%
2,2'-Bipyridyl
Hazard: Irritant


Formula: C₁₀H₈N₂
MW: 156.19 g/mol
MDL Number: MFCD00006212
CAS Number: 366-18-7
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/84/90/2298490.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty