Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform ≥95%

Supplier: Matrix Scientific
Synonyms: Tris(dibenzylideneacetone)dipalladium(0) chloroform adductTris(dibenylideneacetone)dipalladium-chloroformTris(dibenzylideneacetone)dipalladium-chloroform adduct

076439-5G
102546-194EA 238.46 USD InStock
102546-194
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform ≥95%
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform
Hazard: Irritant


Formula: C₅₁H₄₂O₃Pd₂·CHCl₃
MW: 1035.11 g/mol
Melting Pt: 131…135 °C
MDL Number: MFCD00075479
CAS Number: 52522-40-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty