Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-fluorophenylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific
Synonyms: 4-Amino-3-fluorobenzeneboronic acid pinacol ester4-Amino-3-fluorophenylboronic acid, pinacol ester2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

076514-5G 076514-10G 076514-1G
102546-606EA 496.92 USD InStock
102546-606 102546-604 102546-608
4-Amino-3-fluorophenylboronic acid pinacol ester ≥95%
4-Amino-3-fluorophenylboronic acid pinacol ester
Hazard: Irritant


Formula: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
MDL Number: MFCD09033884
CAS Number: 819058-34-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty