Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Oxocyclohexyl)propanenitrile

Supplier: Matrix Scientific

079862-25G 079862-5G
102833-956EA 320 USD InStock
102833-956 102833-954
3-(2-Oxocyclohexyl)propanenitrile
3-(2-Oxocyclohexyl)propanenitrile
Hazard: Irritant


Formula: C₉H₁₃NO
MDL Number: MFCD00013797
CAS Number: 4594-78-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty