Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl (4-fluorobenzoyl)acetate

Supplier: Matrix Scientific
Synonyms: Methyl 4-fluorobenzoylacetateMethyl 3-(4-Fluorophenyl)-3-oxopropionate3-(4-Fluorophenyl)-3-oxopropionic Acid Methyl Ester(4-Fluorobenzoyl)acetic Acid Methyl Ester

080530-500MG 080530-5G 080530-1G
102669-996EA 193.85 USD InStock
102669-996 102670-000 102669-998
Methyl (4-fluorobenzoyl)acetate
Methyl (4-fluorobenzoyl)acetate
CAS: 63131-29-3
MDL Number: MFCD00000355
Molecular Formula: C10H9FO3
Molecular Weight: 196,18
Hazard: Irritant


Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
MDL Number: MFCD00000355
CAS Number: 63131-29-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty