Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dibromoaniline ≥97%

Supplier: Matrix Scientific

081166-5G
102834-230EA 80 USD InStock
102834-230
2,6-Dibromoaniline ≥97%
2,6-Dibromoaniline
Hazard: Toxic


Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 262…264 °C
Melting Pt: 80…82 °C
MDL Number: MFCD00007638
CAS Number: 608-30-0
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty