Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-chlorobenzenesulfonic acid ≥97%

Supplier: Matrix Scientific

081187-1G 081187-10G
102834-286EA 72.31 USD InStock
102834-286 102834-288
2-Amino-5-chlorobenzenesulfonic acid ≥97%
2-Amino-5-chlorobenzenesulphonic acid
Hazard: Irritant


Formula: C₆H₆ClNO₃S
MDL Number: MFCD00035776
CAS Number: 133-74-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty