Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine ≥97%

Supplier: Matrix Scientific
Synonyms: Benzyl-3-aminopyrrolidine-1-carboxylate1-Benzyloxycarbonyl-3-aminopyrrolidine3-Aminopyrrolidine-1-carboxylic acid benzyl ester1-Z-3-aminopyrrolidine1-Cbz-3-aminopyrrolidine

081415-1G
102834-850EA 383.08 USD InStock
102834-850
(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine ≥97%
(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
Hazard: Irritant


Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
MDL Number: MFCD03001673
CAS Number: 185057-50-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty