Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromoisophthalic acid ≥97%

Supplier: Matrix Scientific

082014-50G
102836-330EA 849.23 USD InStock
102836-330
4-Bromoisophthalic acid ≥97%
4-Bromoisophthalic acid
Hazard: Irritant


Formula: C₈H₅BrO₄
MW: 245.03 g/mol
MDL Number: MFCD00002404
CAS Number: 6939-93-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty