Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluic acid ≥98%

Supplier: Matrix Scientific
Synonyms: 4-Toluic acid4-Methylbenzoic acid

082015-50G 082015-10G
102836-332EA 40 USD InStock
102836-332 102836-334
p-Toluic acid ≥98%
p-Toluic acid
Hazard: Irritant


Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 274 °C
Melting Pt: 178…182 °C
Density: 1.06
Flash Pt: 181 °C
MDL Number: MFCD00002565
CAS Number: 99-94-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty