Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-3,5-dibromoanthranilic acid ≥97%

Supplier: Matrix Scientific
Synonyms: 2-Amino-6-chloro-3,5-dibromobenzoic acid

082093-1G
102836-532EA 241.54 USD InStock
102836-532
6-Chloro-3,5-dibromoanthranilic acid ≥97%
6-Chloro-3,5-dibromoanthranilic acid
Hazard: Irritant


Formula: C₇H₄Br₂ClNO₂
MW: 329.37 g/mol
CAS Number: 143769-25-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty