Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-histidine monohydrate ≥97%

Supplier: Matrix Scientific
Synonyms: Ac-His-OH·H2OAc-His-OH.H2O

083053-1G 083053-10G
102838-792EA 72.31 USD InStock
102838-792 102838-794
N-Acetyl-L-histidine monohydrate ≥97%
N-Acetyl-L-histidine monohydrate
Purity: 97%
CAS: 39145-52-3
MDL Number: MFCD00149320
Molecular Formula: C8H13N3O4
Molecular Weight: 215,21
Hazard: Irritant


Formula: C₈H₁₁N₃O₃·H₂O
MW: 215.2 g/mol
MDL Number: MFCD00149320
CAS Number: 39145-52-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty