Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bzl-Gly-OH ≥97%

Supplier: Matrix Scientific
Synonyms: 2-(Benzylamino)acetic acid

083066-50G 083066-10G 083066-1G
102838-836EA 1090.77 USD InStock
102838-836 102838-832 102838-834
Bzl-Gly-OH ≥97%
Bzl-Gly-OH
Hazard: Irritant


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00190772
CAS Number: 17136-36-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty