Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Bromomethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

084324-1G
102842-100EA 553.85 USD InStock
102842-100
4-(Bromomethyl)benzaldehyde ≥97%
4-(Bromomethyl)benzaldehyde
Hazard: Irritant


Formula: C₈H₇BrO
MDL Number: MFCD08271963
CAS Number: 51359-78-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty