Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropyl-4-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific
Synonyms: Cyclopropyl(4-methoxyphenyl)methanone

084615-10G 084615-1G 084615-50G
102842-898EA 329.23 USD InStock
102842-898 102842-894 102842-896
Cyclopropyl-4-methoxyphenyl ketone ≥97%
Cyclopropyl 4-methoxyphenyl ketone
Hazard: Irritant


Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 41…42 °C
MDL Number: MFCD00001296
CAS Number: 7152-03-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty