Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-2-nitrobenzonitrile ≥97%

Supplier: Matrix Scientific

084685-1G 084685-10G
102843-090EA 1421.54 USD InStock
102843-090 102843-088
5-Fluoro-2-nitrobenzonitrile ≥97%
5-Fluoro-2-nitrobenzonitrile
Hazard: Irritant


Formula: C₇H₃FN₂O₂
MDL Number: MFCD01632196
CAS Number: 50594-78-0
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty