Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥97%

Supplier: Matrix Scientific
Synonyms: DTNB5,5'-Disulfanediylbis(2-nitrobenzoic acid)2,2'-Dinitro-5,5'-dithiodibenzoic acidEllman's reagent3,3'-Dithiobis(6-nitrobenzoic acid)5,5'-Disulphanediylbis(2-nitrobenzoic acid)

084824-50G
102843-490EA 849.23 USD InStock
102843-490
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥97%
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
Hazard: Irritant


Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
MDL Number: MFCD00007140
CAS Number: 69-78-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty