Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylacetyl chloride ≥95%

Supplier: Matrix Scientific
Synonyms: 2,2-Diphenylacetyl chloride

083101-1G 083101-10G
102838-918EA 127.69 USD InStock
102838-918 102838-920
Diphenylacetyl chloride ≥95%
Diphenylacetyl chloride
Hazard: Irritant


Formula: C₁₄H₁₁ClO
MW: 230.69 g/mol
MDL Number: MFCD00013655
CAS Number: 1871-76-7
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty