Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Alanine ethyl ester hydrochloride ≥97%

Supplier: Matrix Scientific
Synonyms: L-Ethyl alanate hydrochlorideH-Ala-OEt.HClEthyl (S)-2-Aminopropionate Hydrochloride(S)-2-Aminopropionic Acid Ethyl Ester Hydrochloride

083737-50G 083737-10G 083737-1G
102840-590EA 193.85 USD InStock
102840-590 102840-592 102840-588
L-Alanine ethyl ester hydrochloride ≥97%
L-Alanine ethyl ester hydrochloride
Purity: 97%
CAS: 1115-59-9
MDL Number: MFCD00063662
Molecular Formula: C5H12ClNO2
Molecular Weight: 153,61
Hazard: Irritant


Formula: C₅H₁₁NO₂
MW: 153.61 g/mol
MDL Number: MFCD00063662
CAS Number: 1115-59-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty