Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-iodonicotinonitrile ≥97%

Supplier: Matrix Scientific

085190-1G
102844-424EA 949.23 USD InStock
102844-424
2-Amino-5-iodonicotinonitrile ≥97%
2-Amino-5-iodonicotinonitrile
Hazard: Irritant


Formula: C₆H₄IN₃
MDL Number: MFCD20040120
CAS Number: 1347815-41-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty