Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Butylcyclohexanol ≥97%

Supplier: Matrix Scientific
Synonyms: 1-n-Butylcyclohexanol

085804-10G 085804-1G
102846-030EA 349.23 USD InStock
102846-030 102846-028
1-Butylcyclohexanol ≥97%
1-Butylcyclohexanol
Hazard: Irritant


Formula: C₁₀H₂₀O
MW: 156.27 g/mol
MDL Number: MFCD00021404
CAS Number: 5445-30-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty