Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sucralose ≥97%

Supplier: Matrix Scientific
Synonyms: 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside(2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol

085854-1G
102846-204EA 383.08 USD InStock
102846-204
Sucralose ≥97%
Sucralose
Sucralose
Purity: 97%
CAS: 56038-13-2
MDL Number: MFCD11100146
Molecular Formula: C12H19Cl3O8
Molecular Weight: 397,64
Hazard: Irritant


Formula: C₁₂H₁₉Cl₃O₈
MW: 397.63 g/mol
MDL Number: MFCD03648615
CAS Number: 56038-13-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty