Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetamidine hydrochloride ≥90%

Supplier: Matrix Scientific

086111-50G
102846-974EA 40 USD InStock
102846-974
Acetamidine hydrochloride ≥90%
Acetamidine hydrochloride
Hazard: Irritant/Hygroscopic


Formula: C₂H₇ClN₂
Melting Pt: 168…175 °C
MDL Number: MFCD00013016
CAS Number: 124-42-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty