Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodo-2,5-xylenol ≥97%

Supplier: Matrix Scientific
Synonyms: 5-Hydroxy-2-iodo-4-xylene4-Iodo-2,5-dimethylphenol2,5-Dimethyl-4-iodophenol2-Hydroxy-5-iodo-4-xylene

113740-1G 113740-10G
102892-722EA 350.77 USD InStock
102892-722 102892-724
4-Iodo-2,5-xylenol ≥97%
4-Iodo-2,5-xylenol
Hazard: Irritant


Formula: C₈H₉IO
MW: 248.06 g/mol
MDL Number: MFCD00219922
CAS Number: 114971-53-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty