Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-(pentafluorothio)aniline ≥97%

Supplier: Matrix Scientific
Synonyms: 4-Amino-3-chlorophenylsulphur pentafluoride(4-Amino-3-chlorophenyl)pentafluorosulphur4-Amino-3-chlorophenylsulfur pentafluoride(4-Amino-3-chlorophenyl)pentafluorosulfur

114358-1G
102893-864EA 690.77 USD InStock
102893-864
2-Chloro-4-(pentafluorothio)aniline ≥97%
2-Chloro-4-(pentafluorothio)aniline
Hazard: Irritant


Formula: C₆H₅ClF₅NS
MW: 253.620 g/mol
CAS Number: 165114-85-2
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty