Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methoxy-m-cresol ≥97%

Supplier: Matrix Scientific
Synonyms: 3-Hydroxy-5-methylanisol1-Hydroxy-3-methoxy-5-methylbenzene3-Hydroxy-5-methoxytoluene3-Methoxy-5-methylphenol

114170-1G
102893-494EA 86.15 USD InStock
102893-494
5-Methoxy-m-cresol ≥97%
5-Methoxy-m-cresol
Hazard: Irritant


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00059261
CAS Number: 3209-13-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty