Would you like to log in again?
PIG® Grippy® Traffic Mat® Rug, PIG®
Supplier: New Pig
MAT3223
89508-230EA 467.71 USD InStock
89508-230
PIG® Grippy® Traffic Mat® Rug, PIG®
Spill Control Materials
This durable absorbent mat is the only one that stays put no matter what.
  • Holds tight but peels up easily
  • No bunching, rippling, flipping or shifting
  • Heat-fused and needle-punched for incredible durability
Our most durable absorbent mat is the only one that stays put no matter what® - even under forklift traffic. Full-coverage Grippy® adhesive backing holds tight but peels up easily. No bunching, rippling, flipping or shifting; lays flat and stays flat to help eliminate trips, slips and falls. Heat-fused and needle-punched for incredible durability; won't rip, shred or fray under constant walk-on or wheeled traffic, even from forklifts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty