Would you like to log in again?
MPure Forensic DNA Extraction Kit , MP Biomedicals, LLC
Supplier: MP Biomedicals
MPure™
117022110 117022110
10204-738EA 605.84 USD InStock
10204-738 CA76046-206
MPure Forensic DNA Extraction Kit , MP Biomedicals, LLC
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MPure Forensic DNA Extraction Kit is used with the MPure-12 Nucleic Acid Purification System for the extraction and purification of genomic DNA from forensic samples.
  • Extract and isolate genomic DNA from forensic samples
  • Completely automated purification protocol
  • Use with MPure-12 System
  • Run in as little as 40 minutes
  • DNA is ready for immediate use
The kit contains everything needed for sample preparation. However, for tough samples such as swipes and swabs, cigarette butts and bodily fluid stained fabrics or other solid matrices, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time. DNA extracted using the MPure Forensic DNA Extraction Kit is ready for immediate use and is compatible for quantitation using the Quantifiler™ Human, Quantifiler™ Y Human Male, Quantifiler™ Duo DNA Quantification Kits and Investigator™Quantiplex Kit, and for use in STR amplification using the AmpFISTR™ PCR Amplification kits as well as other downsteam applications.
Capacity. Chewing Gum - 40 mg, Cigarette butts - 1 cm2 paper from the end of the cigarette or filter, Clotted/ dried blood - 20 µL, Forensic Surface and Contact Swabs, Hair root - 0.5–1 cm, Human tissues - 40mg, Saliva - 50µL, Sperm Stains - 5-10 µL or 1 cm2 of the forensic sample
Packaging: Contains Lysis Buffer 2 = 1000 µL, Binding Buffer 1 = 1000 µL, Magnetic Bead Solution = 800 µL, Washing Buffer 1 = 1000 µL, Washing Buffer 2 = 1000 µL, Washing Buffer 3 = 1000 µL, Elution Buffer 1 = 1000 µL, Elution Buffer 2 = 1000 µL
Caution: Store at room temperature (15-25°C). Do not freeze the reagent cartridges. The Kits are stable for 12 months under these conditions. Store the purified DNA at 4°C (short-term) or aliquot and store at -70°C (long-term) before performing the downstream analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty