Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Viral/Pathogen Nucleic Acid Extraction Kit A, MP Biomedicals
Supplier: MP Biomedicals
117022120 117022120
10204-740EA 628.51 USD InStock
CA76046-208 10204-740
Viral/Pathogen Nucleic Acid Extraction Kit A, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MPurify your Nucleic Acid: Obtain Superior Yield, Exceptional Purity, and Automated Performance in a Flexible, Convenient and Affordable Manner!
  • Extract viral and bacterial DNA/RNA from cell-free samples
  • Serum, plasma and other cell-free body fluids
  • Use with MPure-12 System
  • Run in as little as 40 minutes
  • Nucleic Acid is ready for immediate use
The kit contains everything needed for sample preparation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty